Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMMPRIN/CD147 Mouse anti-Human, Clone: 109403, R&D Systems™

Mouse Monoclonal Antibody

$125.00 - $839.00

Specifications

Antigen EMMPRIN/CD147
Immunogen Mouse myeloma cell line NS0-derived recombinant human EMMPRIN/CD147 Ala22-His205 Accession # Q54A51
Clone 109403
Applications Western Blot, ELISA, Immunoassay
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB972SP
View Documents
R&D Systems
MAB972SP
25 ug
Each for $125.00
Add to cart
 
MAB972100
View Documents
R&D Systems
MAB972100
100 ug
Each for $356.00
Add to cart
 
MAB972500
View Documents
R&D Systems
MAB972500
500 ug
Each for $839.00
Add to cart
 
Description

Description

EMMPRIN/CD147 Monoclonal antibody MAB972 Specifically detects EMMPRIN/CD147 in Human samples. It is validated for Western Blotting
Specifications

Specifications

EMMPRIN/CD147
109403
Unconjugated
Q54A51
Monoclonal
Primary
Detects human EMMPRIN/CD147 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse EMMPRIN is observed.
Protein A or G purified from hybridoma culture supernatant
Mouse myeloma cell line NS0-derived recombinant human EMMPRIN/CD147 Ala22-His205 Accession # Q54A51
Western Blot, ELISA, Immunoassay
Purified
Mouse
IgG2b
682
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit