Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMP Mouse anti-Human, Mouse, Rat, Clone: 730340, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524344

Catalog No. PIMA524344


Add to cart

Description

Description

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

This gene product mediates the attachment of erythroblasts to macrophages. This attachment promotes terminal maturation and enucleation of erythroblasts, presumably by suppressing apoptosis. This protein is an integral membrane protein with the N-terminus on the extracellular side and the C-terminus on the cytoplasmic side of the cell. Two immunologically related isoforms of erythroblast macrophage protein with apparent molecular weights of 33 kD and 36 kD were detected in macrophage membranes, this gene encodes the larger isoform. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.
Specifications

Specifications

EMP
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q7L5Y9, Q4VC33, Q5RKJ1
EMLP, EMP, HLC-10, PIG5, cell proliferation-inducing gene 5 protein, erythroblast macrophage protein, human lung cancer oncogene 10 protein, lung cancer-related protein 10, proliferation-inducing gene 5
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10296, 298982, 59003
Immunocytochemistry, Immunofluorescence, Western Blot
730340
Unconjugated
MAEA
Lyophilized
MAEA
E. coli-derived recombit human EMP/MAEA Met12-Ser66
Protein A/G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit