Learn More
EMR2 (CD312) Mouse anti-Human, Alexa Fluor 647, Clone: 494025, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564342
Catalog No. BDB564342
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The 494025 monoclonal antibody specifically binds to EMR2 (Epidermal Growth Factor Module-containing Mucin-like Receptor 2) glycoprotein which is also known as CD312. EMR2 belongs to the Epidermal Growth Factor-seven Span Transmembrane (EFG-TM7) family. EMR2 is expressed by activated lymphocytes and myeloid cells. CD312 can bind to the glycosaminoglycans, dermatan sulfate and chondroitin sulfate. Upon cellular activation, CD312 expression increases and localizes to the leading edge of neutrophils and regulates their adhesion, migration and production of superoxide anion. CD312 expression increases as monocytes differentiate into macrophages but decreases upon differentiation into dendritic cells. EMR2 may also mediate some interactions between activated B cells that express chondroitin sulfate and activated dendritic cells, macrophages and T cells.
Specifications
Specifications
EMR2 (CD312)
494025
CD312; EGF-like module-containing mucin-like hormone receptor-like 2
Affinity Purified
Human EMR2 Recombinant Protein
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG2b, κ
RUO
Primary
Human