Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENDOG Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513513

Catalog No. PIPA513513

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

ENDOG is a nuclear encoded endonuclease that is localized in the mitochondrion. The protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA.
Specifications

Specifications

ENDOG
Polyclonal
PBS with 0.09% sodium azide
Q14249, O08600
Endonuclease G, mitochondrial; Endo G; ENDOG
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
13804, 2021
Flow Cytometry, Western Blot
Unconjugated
ENDOG
Liquid
ENDOG
KLH conjugated synthetic peptide between 188-217 amino acids from the C-terminal region of human ENDOG
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit