Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Endophilin A1/SH3GL2, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP185521
Description
Endophilin A1/SH3GL2 Polyclonal antibody specifically detects Endophilin A1/SH3GL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Endophilin A1/SH3GL2 | |
0.05mg/mL | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Affinity Purified | |
SH3GL2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ALVQAQLEYHKQAVQILQQVTVRLEERIRQASSQPRREYQPKPRMSLEFPTGDSTQPNGGLSHTGTPKPSGVQMDQPCCRALYDFEPENEGELG | |
Immunogen affinity purified | |
RUO | |
Primary | |
6456 | |
Human |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CNSA2FLJ25015, EEN-B1bA335L15.1 (SH3-domain GRB2-like 2), Endophilin-1, endophilin-A1, FLJ20276, SH3 domain protein 2A, SH3 domain-containing GRB2-like protein 2, SH3D2AEndophilin A1 BAR domain, SH3-domain GRB2-like 2, SH3P4endophilin-1 | |
Rabbit | |
IgG | |
0.1 ml | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only