Learn More
Endosialin Mouse anti-Human, Alexa Fluor 647, Clone: B1/35, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564994
Catalog No. BDB564994
Add to cart
Description
Description
The B1/35 monoclonal antibody specifically binds to CD248 which is also known as Endosialin, Tumor endothelial marker 1 (TEM1), and CD164 sialomucin-like 1 (CD164L1). CD248 is a heavily glycosylated, single-pass type I transmembrane protein. CD248 belongs to the Group XIV C-Type lectin family that includes CD93 and CD141/thrombomodulin. It is expressed on pericytes and stromal fibroblasts. Although the exact functions of CD248 remain to be determined, its expression is associated with angiogenesis and lymphoid tissue organization during development, as well as, with postnatal inflammation and tumor development and growth.
Specifications
Specifications
Endosialin
B1/35
Endosialin; TEM1; CD164L1
Affinity purified
Diploid human fibroblasts
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry, Immunohistochemistry, Western Blot
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human