Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endostatin, Polyclonal, Invitrogen™

Catalog No. PIPA130191
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA130191 50 μg
1 options

Catalog No. PIPA130191

Supplier: Thermo Scientific PA130191

Goat Polyclonal Antibody

Endostatin is a 20-kDa proteolytic COOH-terminal fragment derived from collagen XVIII. Endostatin reduces endothelial cell proliferation and acts as a potent inhibitor of angiogenesis and tumor growth.

PA1-30191 detects Endostatin from mouse samples.PA1-30191 has been successfully used in immunohistochemistry (frozen tissue), Western blot applications.The PA1-30191 immunogen is: Recombinant full length Endostatin expressed in insect cell line Sf21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endostatin
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.1mg/mL
Conjugate Unconjugated
Gene Col18a1
Gene Accession No. P39061
Gene Alias Alpha 1 Collagen type 18 (XVIII)(COL18A1), COL18A1, Endostatin, KNO
Gene Symbols COL18A1
Host Species Goat
Immunogen Recombinant full length Endostatin expressed in insect cell line Sf21.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12822
Target Species Mouse
Content And Storage Store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.