Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RECA-1 Mouse anti-Rat, Clone: HIS-52, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135781

Catalog No. 01-672-085


Add to cart

Description

Description

RECA-1 is a cell surface antigen which is expressed by all rat endothelial cells. The endothelium is the thin layer of cells that line the interior surface of blood vessels, forming an interface between circulating blood in the lumen and the rest of the vessel wall. Endothelial cells line the entire circulatory system, from the heart to the smallest capillary.
Specifications

Specifications

RECA-1
Monoclonal
Unconjugated
Liquid
Mouse
IgG1
RUO
Antibody
Monoclonal
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
HIS-52
tissue culture supernatant diluted in PBS with 0.1% sodium azide
Endothelial cells; Endothelium Cell Antigen-1; Endothelial cell marker; Pan endothelial cells
Peripheral and mesenteric lymph nodes isolate.
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit