Learn More
LIPG Rabbit anti-Human, DyLight® 488, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA522913
Catalog No. PIPA522913
Add to cart
Description
Description
This antibody does not react with mouse endothelial lipase.
LIPG (Lipase G, Endohelial type) is an endothelial lipase which is a novel member of the lipoprotein lipase gene family. LIPG is synthesized by endothelial cells and functions at the site of synthesis. LIPG most likely plays a physiological role in modulating lipoprotein metabolism. Overexpressed LIPG has been shown to reduce HDL cholesterol levels, whereas an LIPG deficiency appears to be effective in elevating HDL cholesterol levels in animals. LIPG has substantial phospholipase activity and may be involved in lipoprotein metabolism and vascular biology. Also, LIPG is designated a member of the TG lipase family by its sequence and characteristic lid region which provides substrate specificity for enzymes of the TG lipase family. LIPG is a carboxyl esterase that hydrolyzes high density lipoproteins, emulsifies triglycerides, is essential for the efficient digestion of dietary fats and can bind to heparin.
Specifications
Specifications
LIPG
Polyclonal
DyLight™ 488
LIPG
Liquid
LIPG
A C-terminal synthetic peptide made to the human endothelial lipase protein sequence.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
sodium borate with no preservative
Q9Y5X9
UNQ387/PRO719, EDL, EL, PRO719, endothelial cell-derived lipase, endothelial lipase, lipoprotein lipase H
Rabbit
IgG
100 μL
4° C, store in dark
Primary
9388