Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Endothelial Lipase, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $361.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A C-terminal synthetic peptide made to human Endothelial Lipase [UniProt# Q9Y5X9]
Antigen Endothelial Lipase
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB400111SS
View Documents
Novus Biologicals
NB400111SS
0.025 ml
Each for $119.60
Add to cart
 
NB400111
View Documents
Novus Biologicals
NB400111
0.1 ml
Each for $361.50
Add to cart
 
Description

Description

Endothelial Lipase Polyclonal antibody specifically detects Endothelial Lipase in Human, Mouse (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications

Specifications

Rabbit
A C-terminal synthetic peptide made to human Endothelial Lipase [UniProt# Q9Y5X9]
Polyclonal
Primary
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Q9Y5X9
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
IgG
Endothelial Lipase
Affinity Purified
RUO
Human. Predicted to react with monkey based on 100% sequence homology. Does not appear to recognize mouse.
9388
1 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit