Learn More
anti-Endothelin-1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP133613
Catalog No. NBP133613
Add to cart
Description
Description
Endothelin-1 Polyclonal antibody specifically detects Endothelin-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Endothelin-1
0.52mg/mL
Western Blot 1:500-1:10000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin
Affinity Purified
EDN1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Endothelin 1. The exact sequence is proprietary.
24 kDa
0.1 ml
Cell Cycle and Replication
Primary
1906
Human
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.