Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelin B Receptor Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA120534

Catalog No. PA120534


Add to cart

Description

Description

PA1-20534 detects Endothelin B Receptor in human samples. PA1-20534 has been successfully used in immunohistochemistry (paraffin) procedures. The PA1-20534 immunogen is a synthetic peptide corresponding to the N-terminal extracellular portion of human Endothelin B Receptor.

Receptor B for endothelin 1. Mediates its action by association with G proteins that activate a phosphatidylinositol calcium second messenger system.
Specifications

Specifications

Endothelin B Receptor
Polyclonal
Unconjugated
EDNRB
Liquid
EDNRB
Synthetic peptide corresponding to the N-terminal extracellular portion of human Endothelin B Receptor.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.01% sodium azide; pH 7.7
P24530
RP11-318G21.1, ABCDS, ETB, ETBR, ETRB, HSCR, HSCR2
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1910
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit