Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelin B Receptor Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527173

Catalog No. PIPA527173


Add to cart

Description

Description

PA5-27173 targets Endothelin B Receptor in Western Blot and Immunohistochemistry - Paraffin applications and shows reactivity with Human, mouse and Rat samples. The PA5-27173 immunogen is a synthetic peptide corresponding to a region within amino acids 8 and 111 of Endothelin B Receptor. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
Specifications

Specifications

Endothelin B Receptor
Polyclonal
Unconjugated
EDNRB
Liquid
EDNRB
Synthetic peptide corresponding to a region within amino acids 8 and 111 of Endothelin B Receptor
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7
P24530, P48302, P21451
Ednra, Etb, ET-B, ET-BR, endothelin B receptor, endothelin receptor non-selective type, endothelin-B receptor
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
13618, 1910, 50672
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit