Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD105, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127884

Catalog No. PIPA127884

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Major glycoprotein of vascular endothelium. May play a critical role in the binding of endothelial cells to integrins and/or other RGD receptors. Endoglin is a homodimeric transmembrane glycoprotein highly expressed by endothelial cells. It is a component

PA1-27884 detects ENG from human samples.PA1-27884 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27776 immunogen is a synthetic peptide corresponding to human Endoglin.
Specifications

Specifications

CD105
Polyclonal
PBS with no preservative
P17813
CD105, END, HHT1, ORW, ORW1, CD105 antigen, ENG
Chicken
IgY
50μg
Store at -20°C
Primary
2022
Western Blot
Unconjugated
ENG
Purified
ENG
Synthetic peptide corresponding to human Endoglin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit