Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENO1, Polyclonal, Invitrogen™

Catalog No. PIPA127948
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127948 50 μg
1 options

Catalog No. PIPA127948

Supplier: Thermo Scientific PA127948

Chicken Polyclonal Antibody

Multifunctional enzyme that. as well as its role in glycolysis. plays a part in various processes such as growth control. hypoxia tolerance and allergic responses. May also function in the intravascular and pericellular fibrinolytic system due to its abil

PA1-27948 detects ENO1 from human, mouse, and rat samples.PA1-27948 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27931 immunogen is a synthetic peptide corresponding to human Alpha-enolase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENO1
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene ENO1
Gene Accession No. P06733, P17182, P04764
Gene Alias ENO1L1, MBP-1, MPB1, NNE, PPH
Gene Symbols ENO1
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Alpha-enolase.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2023
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.