Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENO1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513459

Catalog No. PIPA513459


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken, non-human primate, rat and Xenopus based on sequence homology.

ENO1 is one of three enolase isoenzymes found in mammals; it is alpha-enolase, a homodimeric soluble enzyme, and also a shorter monomeric structural lens protein, tau-crystallin. The two proteins are made from the same message. The full length protein, the isoenzyme, is found in the cytoplasm. The shorter protein is produced from an alternative translation start, is localized to the nucleus, and has been found to bind to an element in the c-myc promoter.
Specifications

Specifications

ENO1
Polyclonal
PBS with 0.09% sodium azide
P06733, P17182
Alpha-enolase; 2-phospho-D-glycerate hydro-lyase; Non-neural enolase; NNE; Enolase 1; Phosphopyruvate hydratase; C-myc promoter-binding protein; MBP-1; MPB-1; Plasminogen-binding protein; MPB1; MBPB1; ENO1L1; ENO1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
13806, 2023
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ENO1
Liquid
ENO1
KLH conjugated synthetic peptide between 178-205 amino acids from the central region of human ENO1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit