Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Enolase Mouse, Unlabeled, Clone: 3, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611197

 View more versions of this product

Catalog No. BDB611197


Add to cart

Description

Description

Enolases are glycolytic enzymes that interconvert 2-phosphoglycerate to phosphoenolpyruvate. They exist as three structurally related but genetically distinct isoforms. The active form of the enzyme is a homodimer formed from one of three subunits (α, β, and γ) that are encoded by distinct genes. The expression of each enolase gene is regulated in a tissue- and developmental- specific manner. While α-enolase has been reported to be expressed in nearly all embryonic and adult tissues, a developmental switch occurs between α and γ isoforms in cells of neuronal origin and between α- and β-enolase in developing skeletal muscle and heart. Early in embryogenesis, β-enolase reportedly is expressed at low levels in skeletal primary fibers. Increases in the expression of β-enolase are detected at the fetal stage of development and after birth. Levels of β-enolase are further increased in adult fast-twitch fibers and with terminal differentiation. In addition, β-enolase expression is regulated during hypoxia via the modulation of Sp1/Sp3 transcription factors levels. Thus, β-enolase is a muscle specific enolase that is thought to be essential for proper development and differentiation of myocytes.

Host Species: Mouse
Clone: 3
Isotype: IgG2a
Species Reactivity: Rat
Immunogen: Human ENO-3 aa. 248-363
Formula Weight [Chemical]: 47kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

β-Enolase
3
Unconjugated
Murine
IgG2a
RUO
Primary
Canine, Human, Rat
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Human ENO-3 aa. 248-363
150μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.