Learn More
Entamoeba histolytica Mouse anti-Protozoa, Clone: EH123.5, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110656
Catalog No. 01-671-587
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10656 detects Entamoeba histolytica. MA1-10656 has been successfully used in ELISA applications. The MA1-10656 immunogen is Entamoeba histolytica HK-9.
Specifications
Specifications
Entamoeba histolytica
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
E. histolytica
Entamoeba histolytica HK-9.
Protein G
RUO
Antibody
Monoclonal
ELISA
EH123.5
Unconjugated
Liquid
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Protozoa