Learn More
Entamoeba histolytica Mouse anti-Protozoa, Clone: EH304.6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110658
Catalog No. 01-671-589
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10658 detects Entamoeba histolytica. MA1-10658 has been successfully used in ELISA applications. The MA1-10658 immunogen is Entamoeba histolytica HK-9.
Specifications
Specifications
Entamoeba histolytica
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
E. histolytica
Entamoeba histolytica HK-9.
Protein G
RUO
Antibody
Monoclonal
ELISA
EH304.6
Unconjugated
Liquid
Mouse
IgG3
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Protozoa