Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENTPD6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521393

Catalog No. PIPA521393


Add to cart

Description

Description

PA5-21393 targets ENTPD6(CD39L2) in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-21393 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 114 and 285 of ENTPD6. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

ENTPD6
Polyclonal
Unconjugated
ENTPD6
Liquid
ENTPD6
Recombit protein fragment contain a sequence corresponding to a region within amino acids 114 and 285 of ENTPD6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O75354
RP4-738P15.5, CD39L2, IL-6SAG, IL6ST2, NTPDase-6, dJ738P15.3, CD39 antigen-like 2, CD39-like 2, HCV-E2 binding protein 1, NTPDase 6, ectonucleoside triphosphate diphosphohydrolase 6, ectonucleoside triphosphate diphosphohydrolase 6 (putative function), interleukin 6 signal transducer-2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
955
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit