Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENY2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525296

Catalog No. PIPA525296

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Specifications

Specifications

ENY2
Polyclonal
PBS with 0.09% sodium azide
Q9NPA8, Q9JIX0
e(y)2, enhancer of yellow 2 transcription factor homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
223527, 56943
Western Blot
Unconjugated
ENY2
Liquid
ENY2
KLH conjugated synthetic peptide between 2-31 amino acids from the N-terminal region of human ENY2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit