Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eosinophil derived neurotoxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP191872
Description
Eosinophil derived neurotoxin Polyclonal specifically detects Eosinophil derived neurotoxin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Eosinophil derived neurotoxin | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
P10153 | |
RNASE2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:QFTWAQWFETQHINMTSQQCTNAMQV | |
0.1 mL | |
Protein Phosphatase | |
6036 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EC 3.1.27.5, EDNRNase 2, Eosinophil-derived neurotoxin, non-secretory ribonuclease, Ribonuclease 2, Ribonuclease US, ribonuclease, RNase A family, 2 (liver, eosinophil-derived neurotoxin), RNS2RNase UpI-2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction