Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eotaxin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112544

Catalog No. 01-674-787


Add to cart

Description

Description

A suggested positive control for this product is recombit Eotaxin protein.

Eotaxin is a 74-amino acid, eosinophil-chemotactic CC chemokine originally found in bronchoalveolar lavage fluid from allergic inflammatory subjects. It is involved in regulating the recruitment and activation of inflammatory leukocytes, particularly eosinophils. Eotaxin binds and activates the CCR3 chemokine receptor, and may play a fundamental role in the development of allergic responses.
Specifications

Specifications

Eotaxin
Polyclonal
PBS with 0.02% sodium azide
P51671
Small inducible cytokine A11, CCL11, Eosinophil chemotactic protein
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
6356
Western Blot
Unconjugated
CCL11
Liquid
CCL11
Synthetic peptide corresponding to residues 83-97 of human eotaxin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit