Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-2 alpha Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524200

Catalog No. PIPA524200

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.
Specifications

Specifications

HIF-2 alpha
Polyclonal
Unconjugated
EPAS1
Liquid
EPAS1
KLH conjugated synthetic peptide between 124-150 amino acids from the N-terminal region of human EPAS1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q99814
ECYT4, HIF2A, HLF, MOP2, PASD2, bHLHe73, EPAS-1, HIF-1-alpha-like factor, HIF-1alpha-like factor, HIF-2-alpha, HIF2-alpha, PAS domain-containing protein 2, basic-helix-loop-helix-PAS protein MOP2, class E basic helix-loop-helix protein 73, endothelial PAS domain-containing protein 1, hypoxia-inducible fa
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2034
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit