Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPB41L3 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11061027G
Description
EPB41L3 Polyclonal specifically detects EPB41L3 in Human, Mouse, Rat, Primate, Xenopus samples. It is validated for Western Blot.Specifications
EPB41L3 | |
Polyclonal | |
Western Blot | |
Q9Y2J2 | |
EPB41L3 | |
A synthetic peptide made to an internal region of human EPB41L3 (within residues 400-500). [Swiss-Prot# Q9Y2J2] | |
0.1 mL | |
Cancer, Cytoskeleton Markers, Signal Transduction, Tumor Suppressors | |
23136 | |
Store at 4C in the dark. |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
4.1BFLJ37633, DAL1DAL-1,, Differentially expressed in adenocarcinoma of the lung protein 1, erythrocyte membrane protein band 4.1-like 3, KIAA0987band 4.1-like protein 3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Primate, Xenopus species | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction