Learn More
EpCAM Mouse anti-Human, PE, Clone: VU-1D9, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15782
Catalog No. A15782
Add to cart
Description
Description
EGP40 is a 40kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. It reportedly distinguishes adenocarcinomas from pleural mesotheliomas.
Specifications
Specifications
EpCAM
Monoclonal
PE
EPCAM
Liquid
EPCAM
Human CD326
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
VU-1D9
PBS with 0.2% BSA and 15mM sodium azide
P16422
Epithelial Specific Antigen, ESA, KSA, M4S1, MK-1, DIAR5, EGP-2, EGP40, KS1/4, MIC18, TROP1, TACSTD1, EPCAM, Epithelial cell adhesion molecule
Mouse
IgG1
100 Tests
4° C
Primary
4072