Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EpCAM Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525358

Catalog No. PIPA525358


Add to cart

Description

Description

EpCAM is a carcinoma-associated antigen and is amember of a family that includes at least two type I membraneproteins. This antigen is expressed on most normal epithelial cellsand gastrointestinal carcinomas and functions as a homotypiccalcium-independent cell adhesion molecule. The antigen is beingused as a target for immunotherapy treatment of human carcinomas.
Specifications

Specifications

EpCAM
Polyclonal
Unconjugated
EPCAM
Liquid
EPCAM
KLH conjugated synthetic peptide between 57-85 amino acids from the N-terminal region of human EpCAM
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P16422
Epithelial Specific Antigen, ESA, KSA, M4S1, MK-1, DIAR5, EGP-2, EGP40, KS1/4, MIC18, TROP1, TACSTD1, EPCAM, Epithelial cell adhesion molecule
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4072
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit