Learn More
anti-EpCAM/TROP1, Alexa Fluor 488, Clone: 158206, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9601G
Catalog No. FAB9601G
Add to cart
This item is not returnable. View return policy
Description
Description
Applications:
Flow Cytometry
Specifications
Specifications
EpCAM/TROP1
158206
Flow Cytometry 5 uL/10^6 cells
Purified
EPCAM
Mouse myeloma cell line NS0-derived recombinant human EpCAM/TROP-1 Extracellular domain
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
4072
Human
Flow Cytometry
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
Mouse
IgG2b
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human EpCAM/TROP-1 in ELISAs.