anti-EpCAM/TROP1, Polyclonal, Novus Biologicals™

Manufacturer: Novus BiologicalsSupplier Diversity Partner 19300002

 View all options for this product

Catalog No. 19-300-002

  Check Availability
Add to Cart


For Research Use Only


    Antigen EpCAM/TROP1
    Applications ELISA,Western Blotting
    Conjugate Unlabeled
    Cross Reactivity Human
    Format Affinity Purified
    Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
    Gene Accession No. P16422
    Host Species Rabbit
    Immunogen This antibody is specific for the N Terminus Region of the target protein (NM_002354).
    Quantity 0.05mg
    Regulatory Status RUO
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Gene ID (Entrez) 4072
    Test Specificity This product is specific for Human EpCAM.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit