Learn More
MilliporeSigma™ anti-Eph A2/A3/A4/A5, phospho (Tyr 596/602), Polyclonal, (Chemicon) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB2250/DEL
Catalog No. 50-173-376
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene is a receptor for ephrin-B family members.
Western Blotting
Specifications
Specifications
Eph A2/A3/A4/A5, phospho (Tyr 596/602)
Unconjugated
Rabbit
100μL
Neuroscience
Polyclonal
0.22mg/mL
Affinity Purified
Synthetic linear peptide.
RUO
Primary
Human