Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130295

Catalog No. PIPA130295

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

PA1-30295 detects EphA5 from human, and mouse samples. PA1-30295 has been successfully used in immunohistochemistry (frozen tissue), immunohistochemistry (paraffin tissue), and Western blot applications. Positive Control: Mouse brain tissue lysate The PA1-30295 immunogen is a synthetic peptide conjugated to KLH, corresponding to amino acids 957-972 of Human Eph A5. Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20° C or below, avoiding multiple freeze-thaw cycles.

Specifications

Specifications

EpHA5
Polyclonal
Unconjugated
EPHA5
Liquid
EPHA5
Synthetic peptide conjugated to KLH, corresponding to amino acids 957-972 of Human Eph A5.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 0.09% sodium azide
P54756, Q60629
AL-1, AV158822, EFL-5, Ephrin-A5, Epl7, LERK-7, RAGS
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
13839, 2044
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit