Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPHB1, Mouse, MaxPab™ Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human EPHB1 protein.

Manufacturer:  Abnova Corporation H00002047B01

Catalog No. 89-002-679


Add to cart

Description

Description

Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The protein encoded by this gene is a receptor for ephrin-B family members. [provided by RefSeq

Sequence: MALDYLLLLLLASAVAAMEETLMDTRTATAELGWTANPASGWEEVSGYDENLNTIRTYQVCNVFEPNQNNWLLTTFINRRGAHRIYTEMRFTVRDCSSLPNVPGSCKETFNLYYYETDSVIATKKSAFWSEAPYLKVDTIAADESFSQVDFGGRLMKVNTEVRSFGPLTRNGFYLAFQDYGACMSLLSVRVFFKKCPSIVQNFAVFPETMTGAESTSLVIARGTCIPNAEEVDVPIKLYCNGDGEWMVPIGRCTCKPGYEPENSVACKACPAGTFKASQEAEGCSHCPSNSRSPAEASPICTCRTGYYRADFDPPEVACTSVPSGPRNVISIVNETSIILEWHPPRETGGRDDVTYNIICKKCRADRRSCSRCDDNVEFVPRQLGLTECRVSISSLWAHTPYTFDIQAINGVSSKSPFPPQHVSVNITTNQAAPSTVPIMHQVSATMRSITLSWPQPEQPNGIILDYEIRYYEKEHNEFNSSMARSQTNTARIDGLRPGMVYVVQVRARTVAGYGKFSGKMCFQTLTDDDYKSELREQLPLIAGSAAAGVVFVVSLVAISIVCSRKRAYSKEAVYSDKLQHYSTGRGSPGMKIYIDPFTYEDPNEAVREFAKEIDVSFVKIEEVIGAGEFGEVYKGRLKLPGKREIYVAIKTLKAGYSEKQRRDFLSEASIMGQFDHPNIIRLEGVVTKSRPVMIITEFMENGALDSFLRQNDGQFTVIQLVGMLRGIAAGMKYLAEMNYVHRDLAARNILVNSNLVCKVSDFGLSRYLQDDTSDPTYTSSLGGKIPVRWTAPEAIAYRKFTSASDVWSYGIVMWEVMSFGERPYWDMSNQDVINAIEQDYRLPPPMDCPAALHQLMLDCWQKDRNSRPRFAEIVNTLDKMIRNPASLKTVATITAVPSQPLLDRSIPDFTAFTTVDDWLSAIKMVQYRDSFLTAGFTSLQLVTQMTSEDLLRIGITLAGHQKKILNSIHSMRVQISQSPTAMA
Specifications

Specifications

EPHB1
Unconjugated
No additive
NM_004441.3
ELK|EPHT2|FLJ37986|Hek6|NET
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human EPHB1 protein.
EPHB1
Liquid
EPHB1
EPHB1 (NP_004432.1, 1 a.a. ~ 984 a.a) full-length human protein.
RUO
Primary
2047
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit