Learn More
anti-EphB2, APC, Clone: 512012, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Manufacturer:  R&D Systems FAB467A
Catalog No. FAB467A
Add to cart
This item is not returnable. View return policy
Description
Description
EphB2 Monoclonal specifically detects EphB2 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
EphB2
512012
Flow Cytometry 10 uL/10^6 cells
P54763
CAPB, Cek5, Drt, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, Erk, ERKHek5, HEK5, Nuk, PCBC, protein-tyrosine kinase HEK5, Qek2, Renal carcinoma antigen NY-REN-47, Sek3, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5
Rat
IgG2a
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human and mouse EphB2 in direct ELISAs.
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
EPHB2
Mouse myeloma cell line NS0-derived recombinant mouse EphB2 Val27-Lys548 (predicted) Accession # P54763
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
2048
Human, Mouse