Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphB2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514608

Catalog No. PIPA514608


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.
Specifications

Specifications

EphB2
Polyclonal
PBS with 0.09% sodium azide
P29323
Ephrin type-B receptor 2; Tyrosine-protein kinase receptor EPH-3; DRT; Receptor protein-tyrosine kinase HEK5; ERK
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2048
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
EPHB2
Liquid
EPHB2
Recombit human EphB2 protein
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit