Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ephrin-A1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP152103

Catalog No. NBP152103


Add to cart

Description

Description

Ephrin-A1 Polyclonal antibody specifically detects Ephrin-A1 in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

Ephrin-A1
0.5 mg/mL
Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:16000
NP_004419.2
ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
Goat
IgG
0.1 mg
Cancer
Primary
1942
Mouse, Rat
ELISA, Western Blot
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
EFNA1
Peptide with sequence C-KVTVSGKITHSP corresponding to internal region according to NP_004419.2.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize isoform a (NP_004419.2) only.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only