Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ephrin B2, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP149857

Catalog No. NBP149857


Add to cart

Description

Description

Ephrin-B2 Polyclonal antibody specifically detects Ephrin-B2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications

Specifications

Ephrin-B2
0.5 mg/mL
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:16000
P52799
ephrin B2, EPLG5, Htk-L, HTKL, LERK5
Goat
IgG
0.1 mg
Neuroscience, Signal Transduction
Primary
1948
Human, Mouse, Rat
ELISA, Western Blot
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
EFNB2
Peptide with sequence CPKVDSKTVGQYE corresponding to internal region according to NP_004084.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognizes and epitope within the internal region.This antibody is specific to the internal region.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only