Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin B2, Polyclonal, Invitrogen™

Catalog No. PIPA130384
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA130384 50 μg
1 options

Catalog No. PIPA130384

Supplier: Thermo Scientific PA130384

Goat Polyclonal Antibody

Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to

PA1-30384 detects EphB2 from mouse samples.PA1-30384 has been successfully used in ELISA, Western blot applications.The PA1-30384 immunogen is: Recombinant full length protein (Mouse). Expressed in NSO cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin B2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1mg/mL
Conjugate Unconjugated
Gene Efnb2
Gene Accession No. P52800
Gene Alias EFNB2
Gene Symbols EFNB2
Host Species Goat
Immunogen Recombinant full length protein (Mouse). Expressed in NSO cells.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13642
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.