Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSPG3 Mouse anti-Human, Clone: 329707, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524057

Catalog No. PIMA524057


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit mouse DSPG3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

DSPG3
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q99645
Epiphycan, EPYC, PG-Lb, SLRR3B, Dermatan sulfate proteoglycan 3dermatan sulphate proteoglycan 3, DSPG3proteoglycan-lb, epiphycan, PGLB, Pg-Lb, PG-Lb, Proteoglycan-Lb, SLRR3Bepiphycan proteoglycan, Small chondroitin/dermatan sulfate proteoglycan
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1833
Immunocytochemistry, Immunofluorescence, Western Blot
329707
Unconjugated
EPYC
Lyophilized
EPYC
Mouse myeloma cell line NS0-derived recombit human DSPG3 Ala20-Val322
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit