Learn More
anti-epithelial Sodium Channel alpha, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP146603
Catalog No. NBP146603
Add to cart
Description
Description
epithelial Sodium Channel alpha Polyclonal antibody specifically detects epithelial Sodium Channel alpha in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
epithelial Sodium Channel alpha
LYOPH
Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
Purified
SCNN1A
A synthetic peptide from the extracellular domain of human amiloride sensitive sodium channel subunit alpha (SCNN1A, SCNEA, Alpha-ENaC) conjugated to an immunogenic carrier protein was used as the antigen.
IgG purified
RUO
Primary
6337
Reconstitute with 0.5 ml sterilized water.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Sterilized water with No Preservative
alpha ENaC-2, alpha-ENaC, Alpha-NaCH, amiloride-sensitive epithelial sodium channel alpha subunit, amiloride-sensitive sodium channel subunit alpha, amiloride-sensitive sodium channel subunit alpha 2, BESC2, ENaCA, ENaCalpha, Epithelial Na(+) channel subunit alpha, FLJ21883, nasal epithelial sodium channel alpha subunit, Nonvoltage-gated sodium channel 1 subunit alpha, SCNEA, SCNN1, sodium channel, nonvoltage-gated 1 alpha
Rabbit
IgG
0.5 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specific for SCNN1A.
Human