Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPM2A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511517

Catalog No. PIPA511517

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

EPM2A is a dual-specificity phosphatase that associates with polyribosomes. The encoded protein may be involved in the regulation of glycogen metabolism. Mutations have been associated with myoclonic epilepsy of Lafora.
Specifications

Specifications

EPM2A
Polyclonal
PBS with 0.09% sodium azide
B3EWF7
Laforin; Lafora PTPase; LAFPTPase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7957
Immunohistochemistry, Western Blot
Unconjugated
EPM2A
Liquid
EPM2A
KLH conjugated synthetic peptide between 288-317 amino acids from the C-terminal region of human EPM2A
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit