Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-EPM2AIP1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131063

Catalog No. NBP131063


Add to cart

Description

Description

EPM2AIP1 Polyclonal antibody specifically detects EPM2AIP1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

EPM2AIP1
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
EPM2AIP1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human EPM2AIP1. The exact sequence is proprietary.
70 kDa
100 ul
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (83%) and Rat (83%). Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
EPM2A (laforin) interacting protein 1, EPM2A interacting protein 1, EPM2A-interacting protein 1, KIAA0766FLJ11207, Laforin-interacting protein
Rabbit
IgG
Antigen Affinity-purified
RUO
Primary
9852
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only