Learn More
Sino Biological EPOR/Erythropoietin Receptor Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 50031R14050
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EPOR/Erythropoietin Receptor (rM EPOR/Erythropoietin Receptor; Catalog#50031-M08H; NP_034279.3; Met1-Pro249).
Catalog No. NC1477501