Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eps15 Mouse, Unlabeled, Clone: 17, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610807

 View more versions of this product

Catalog No. BDB610807


Add to cart

Description

Description

The eps15 (EGF receptor pathway substrate clone number 15) gene was isolated using an expression cloning method designed for direct isolation of cDNAs encoding substrates of tyrosine kinases. The eps15 gene encodes a 145kDa protein that is tyrosine phosphorylated following EGF receptor activation and this receptor directly phosphorylates purified Eps15 in vitro. Phosphorylation of Eps15 is much more efficient following EGF receptor activation versus erbB2 kinase activation, suggesting the protein is predominantly a part of an EGF receptor signaling pathway. Overexpression of Eps15 causes transformation of NIH 3T3 cells, implying Eps15 may be involved in the control of cell proliferation. Eps15 binds to the SH3 domain of Crk through its proline-rich domain at the C-terminal region. Adaptin α binds to the C-terminus of Eps15 at three different sites ranging from residues 650 to 730. Eps15, Adaptin α and EGF receptor are co-localized at the clathrin-coated pits after receptor activation. Therefore, Eps15 may recruit EGF receptors to the clathrin-coated pits for internalization of the activated receptor.

Immunofluorescence, Immunoprecipitation, Western Blotting

Specifications

Specifications

Eps15
17
Unconjugated
Affinity Purified
Human Eps15 aa. 694-888
150μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.