Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPS15, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA146463

Catalog No. PA146463

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

EPS15 is a ubiquitously expressed protein that is involved in cell growth regulation and may be involved in the regulation of mitogenic signals and control of cell proliferation. Phosphorylation of EPS15 on Tyrosine 849 upon DNA damage is involved in the internalization of EGFR. A chromosomal aberration involving EPS15 is also found in acute leukemias, resulting in a rogue activator protein.

Suggested positive control: antigen standard for EPS15 (transient overexpression lysate), Hela whole cell extract.
Specifications

Specifications

Eps15
Polyclonal
Unconjugated
EPS15
Purified
EPS15
Synthetic peptide corresponding to residues 840-896 of the C-terminal portion of human Eps15. [Swiss-Prot# P42566]
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.75mg/mL
PBS with 0.1% BSA and 0.04% sodium azide
P42566, P42567
Epidermal growth factor receptor substrate 15 , Protein Eps15 , AF-1p protein , EPS15 , AF1P
Rabbit
IgG
100μL
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
2060
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit