Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epstein Barr Virus Nuclear Antigen 1 Goat anti-Virus, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA17497

Catalog No. PA17497


Add to cart

Description

Description

The PA1-7497 antibody detects Epstein-Barr nuclear antigen (EBNA-1). PA1-7497 has been successfully used in ELISA and Western blot applications. Potential applications for PA1-7497 include fluorescence microscopy and immunohistochemistry. The PA1-7497 antibody was raised against recombit EBNA-1.

Epstein-Barr virus (EBV) is a human herpes virus, which is associated with conditions such as Hodgkin ft.s disease and Burkitt ft.s Lymphoma and is the causative agent in mononucleosis in adolescents. EBV latently infects B lymphocytes. Infected B cells express EBV nuclear antigens and latent proteins LMP1, LMP2A and LMP2B. LMP2A forms aggregates in the plasma membranes of B lymphocytes, where it functions as a negative regulator of the Src and Syk protein tyrosine kinases. Studies show that LMP2A blocks B-cell receptor (BCR) signal transduction in EBV immortalized B cells in vitro and may play an important role in maintaining a latent EBV infection within the peripheral blood B cells of infected individuals.EBNA-1 is the only viral protein consistently expressed in all maligcies associated with EBV.
Specifications

Specifications

Epstein Barr Virus Nuclear Antigen 1
Polyclonal
Unconjugated
Liquid
Goat
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Virus
ELISA, Western Blot
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
EBV nuclear antigen 1; EBNA-1
Recombit EBNA-1.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit