Learn More
erbB-3/HER-3 Mouse anti-Human, Mouse, Clone: H3.105.5, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05471
Catalog No. 05-471-MI
Add to cart
Description
Description
Anti-erbB-3/HER-3 Antibody, clone H3.105.5 detects level of erbB-3/HER-3 and has been published and validated for use in immunoprecipitation, NEUT.
Specifications
Specifications
erbB-3/HER-3
H3.105.5
Purified mouse monoclonal IgG1 in buffer containing 10mM PBS, pH 7.4. Frozen solution.
Purified
Murine
IgG1
100μg
Primary
Human, Murine
Immunoprecipitation, Neutralization
Unconjugated
NM_001982
ERBB3, c-erbB-3, c-erbB3, erbB-3, p180-ErbB3, p85-sErbB3, p45-sErbB3, ErbB-3, erbB3-S, MGC88033, HER3, MDA-BF-1
Extracellular domain of recombinant human erbB-3/HER-3 oncoprotein. Clone H3.105.5 Antibody Class IgG1, antibody produced by P3X63Ag8U1 (P3U1) x Balb/c spleen cells. Purified by ammonium sulfate precipitation, Sephacryl S-300 gel filtration and DEAE-52
Protein G purfied
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal