Learn More
erbB-4/HER-4 Mouse anti-Human, Clone: H4.72.8, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05478
Catalog No. 05-478-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This Anti-erbB-4/HER-4 Antibody, clone H4.72.8 is validated for use in flow cytometry, immunoprecipitation, NEUT for the detection of erbB-4/HER-4.
Specifications
Specifications
erbB-4/HER-4
H4.72.8
10mM PBS, pH 7.4
Purified
Murine
IgG2a
100μg
Primary
Human
Flow Cytometry, Immunoprecipitation, Neutralization
Unconjugated
NM_005235
ERBB3, c-erbB-3, c-erbB3, erbB-3, p180-ErbB3, p85-sErbB3, p45-sErbB3, ErbB-3, erbB3-S, MGC88033, HER3, MDA-BF-1
Extracellular domain of recombinant human c-erbB-4/HER-4
Protein A chromatography
−20°C for 24 months.
Monoclonal