Learn More
erbB-4/HER-4 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06572
Catalog No. 06-572-MI
Add to cart
Description
Description
Anti-erbB-4/HER-4 Antibody is an antibody against erbB-4/HER-4 for use in immunoprecipitation and western blotting.
Specifications
Specifications
erbB-4/HER-4
Unconjugated
Purified
Rabbit
IgG
200μg
Primary
NM_005235
Immunoprecipitation, Western Blot
10mM PBS, pH 7.4, with 0.05% sodium azide
ERBB4; p180erbB4; HER4; MGC138404
peptide corresponding to the C-terminus amino acids 1285 to 1308 of human erbB-4/HER-4
Protein A chromatography
−20°C for 24 months
Polyclonal
Human