Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ErbB2/Her2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $293.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Synthetic peptide made to C-terminal portion of human HER2 protein (between residues 1100-1200) [UniProt P04626]
Antigen ErbB2/Her2
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP229624SS
View Documents
Novus Biologicals
NBP229624SS
0.025 mg
Each for $119.60
Add to cart
 
NBP229624
View Documents
Novus Biologicals
NBP229624
0.1 mg
Each for $293.00
Add to cart
 
Description

Description

ErbB2/Her2 Polyclonal antibody specifically detects ErbB2/Her2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Rabbit
Synthetic peptide made to C-terminal portion of human HER2 protein (between residues 1100-1200) [UniProt P04626]
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
PBS with 0.05% Sodium Azide
IgG
ErbB2/Her2
Purified
RUO
2064
1.0 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit