Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
erbB2 (intracellular domain) Mouse anti-Human, Mouse, Clone: 24B5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04291
Description
Anti-erbB2 (intracellular domain) Antibody, clone 24B5 detects level of erbB2 (intracellular domain) and has been published and validated for use in ELISA and western blotting.Specifications
erbB2 (intracellular domain) | |
Monoclonal | |
Unconjugated | |
NM_001005862.1; NM_004448.2 | |
Mouse | |
Subsequent thiophilic adsorption and size exclusion chromatography | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG |
ELISA, Western Blot | |
24B5 | |
lyophilized in 2X PBS containing 0.09% sodium azide, PEG, and sucrose. | |
ERBB2, HER2, HER-2, NGL, herstatin, erbB-2, C-erbB-2, NEU, TKR1, p185erbB2, HER-2/neu | |
KLH-conjugated synthetic peptide encompassing the surrounding amino acids of the intracellular domain in human erbB2 | |
100 μg | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only